Learn More
Aviva Systems Biology Corp Cbfa2t2 Antibody - middle region (ARP37524_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37524_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cbfa2t2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: A430091M07, C330013D05Rik, Cbfa2t2h, MTGR1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-649