Learn More
Aviva Systems Biology Corp CBFA2T2 Antibody - middle region (ARP38559_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38559_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CBFA2T2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CBFA2T2. Alternate names: DKFZp313F2116, EHT, MTGR1, ZMYND3, p85ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56704