Learn More
Aviva Systems Biology Corp CBFA2T2H Antibody - middle region (ARP37525_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37525_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CBFA2T2H. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse CBFA2T2H. Alternate names: MTGR1, Cbfa2t2h, A430091M07, C330013D05RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56378