Learn More
Aviva Systems Biology Corp CBFA2T3 Antibody - N-terminal region (ARP38578_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38578_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CBFA2T3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CBFA2T3. Alternate names: ETO2, MTG16, MTGR2, ZMYND4ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56710