Learn More
BioVendor, LLC Corticosteroid-Binding Globulin Human ELISA, 96-wells, 1/EA
SDP
Supplier:  BioVendor, LLC RD192234200R
Encompass
Corticosteroid-Binding Globulin Human ELISA, 96-wells, 1/EA TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-Heparin, Plasma-CitrateSample Requirements5 ¿l/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2¿8¿C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range3.13¿200 ng/mlLimit of Detection0.01 ng/ml
Catalog No. 50-376-4