Learn More
Medchemexpress LLC CBI1 formic | 461.40 | 100 MG
SDP
Catalog No. 5000392895
Encompass_Preferred
CBI1 formic is a covalent BAX inhibitor that selectively derivatizes BAX at C126. It inhibits BAX activation triggered by ligands or point mutagenesis, blocking t-2-hex lipidation and oligomerization of BAX.
  • Inhibits BAX activation induced by BH3 ligands
  • Blocks t-2-hex lipidation and oligomerization
  • For research use only
Catalog No. 50-003-92895 Supplier Medchemexpress LLC Supplier No. HY161242A100MG
CBI1 formic is a covalent BAX inhibitor that selectively derivatizes BAX at C126. It inhibits BAX activation triggered by ligands or point mutagenesis, blocking t-2-hex lipidation and oligomerization of BAX.
  • Inhibits BAX activation induced by BH3 ligands
  • Blocks t-2-hex lipidation and oligomerization
  • For research use only