Learn More
Medchemexpress LLC CBI1 formic | 461.40 | 5 MG
SDP
Catalog No. 5000391683
Encompass_Preferred
CBI1 formic is a covalent BAX inhibitor. It selectively derivatizes BAX at C126 and inhibits BAX activation by triggering ligands or point mutagenesis. CBI1 formic blocks t-2-hex lipidation and oligomerization of BAX and inhibits BAX activation induced by BH3 ligands, F116A mutagenesis or t-2-hex.
  • Covalent BAX inhibitor
  • Selectively derivatizes BAX at C126
  • Inhibits BAX activation by triggering ligands or point mutagenesis
  • Blocks t-2-hex lipidation and oligomerization of BAX
  • Inhibits BAX activation induced by BH3 ligands, F116A mutagenesis or t-2-hex
Catalog No. 50-003-91683 Supplier Medchemexpress LLC Supplier No. HY161242A5MG
CBI1 formic is a covalent BAX inhibitor. It selectively derivatizes BAX at C126 and inhibits BAX activation by triggering ligands or point mutagenesis. CBI1 formic blocks t-2-hex lipidation and oligomerization of BAX and inhibits BAX activation induced by BH3 ligands, F116A mutagenesis or t-2-hex.
  • Covalent BAX inhibitor
  • Selectively derivatizes BAX at C126
  • Inhibits BAX activation by triggering ligands or point mutagenesis
  • Blocks t-2-hex lipidation and oligomerization of BAX
  • Inhibits BAX activation induced by BH3 ligands, F116A mutagenesis or t-2-hex