Learn More
Medchemexpress LLC CBI1 formic | 461.40 | 50 MG
SDP
Catalog No. 5000392270
Encompass_Preferred
CBI1 formic is a covalent BAX inhibitor that selectively derivatizes BAX at C126. It inhibits BAX activation by triggering ligands or point mutagenesis, and also blocks t-2-hex lipidation and oligomerization of BAX. CBI1 formic is effective in inhibiting BAX activation induced by BH3 ligands, F116A mutagenesis or t-2-hex.
  • Covalent BAX inhibitor
  • Selectively derivatizes BAX at C126
  • Inhibits BAX activation by triggering ligands or point mutagenesis
  • Blocks t-2-hex lipidation and oligomerization of BAX
  • Inhibits BAX activation induced by BH3 ligands, F116A mutagenesis or t-2-hex
  • Available in 50 mg quantity
Catalog No. 50-003-92270 Supplier Medchemexpress LLC Supplier No. HY161242A50MG
CBI1 formic is a covalent BAX inhibitor that selectively derivatizes BAX at C126. It inhibits BAX activation by triggering ligands or point mutagenesis, and also blocks t-2-hex lipidation and oligomerization of BAX. CBI1 formic is effective in inhibiting BAX activation induced by BH3 ligands, F116A mutagenesis or t-2-hex.
  • Covalent BAX inhibitor
  • Selectively derivatizes BAX at C126
  • Inhibits BAX activation by triggering ligands or point mutagenesis
  • Blocks t-2-hex lipidation and oligomerization of BAX
  • Inhibits BAX activation induced by BH3 ligands, F116A mutagenesis or t-2-hex
  • Available in 50 mg quantity