Learn More
Medchemexpress LLC CBI1 formic | 461.40 | C16H21BrN4O3S2 | 10 MG
SDP
Catalog No. 5000288769
Encompass_Preferred
CBI1 formic is a covalent small-molecule inhibitor of BAX that selectively derivatizes the protein at cysteine 126 (C126), preventing BAX activation. Supplied for research use, structural and storage information is provided by the manufacturer.
  • Covalent BAX inhibitor that derivatizes C126.
  • Inhibits BAX activation induced by ligands or point mutations.
  • Offered as a solid with sealed, moisture-free storage recommendations.
  • Molecular formula C16H21BrN4O3S2; molecular weight 461.40.
  • Available in small research pack sizes, including 10 mg.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-88769 Supplier Medchemexpress LLC Supplier No. HY161242A10MG
CBI1 formic is a covalent small-molecule inhibitor of BAX that selectively derivatizes the protein at cysteine 126 (C126), preventing BAX activation. Supplied for research use, structural and storage information is provided by the manufacturer.
  • Covalent BAX inhibitor that derivatizes C126.
  • Inhibits BAX activation induced by ligands or point mutations.
  • Offered as a solid with sealed, moisture-free storage recommendations.
  • Molecular formula C16H21BrN4O3S2; molecular weight 461.40.
  • Available in small research pack sizes, including 10 mg.
  • Intended for research use only; not for human or clinical use.