Learn More
Santa Cruz Biotechnology Cbl-b Antibody (G-1), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC8006
Encompass
Anti-Cbl-b Antibody (G-1) is recommended for detection of Cbl-b of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA.
Catalog No. NC1848929