Learn More
Medchemexpress LLC Cbl-b-IN-1 | 2368841-84-1 | 99.66% | 498.62 | 50 MG
SDP
Catalog No. 5000347973
Encompass_Preferred
Cbl-b-IN-1 (Example 519) is an inhibitor of Cbl-b, with an IC50 of less than 100 nM. It is used in research on immunomodulation. It promotes the secretion of cytokines such as IL-2, IFN-γ, and TNF-α, facilitates T cell activation, and enhances the TCR signaling pathway.
  • Inhibitor of Cbl-b with an IC50 of less than 100 nM
  • Promotes secretion of cytokines (IL-2, IFN-γ, TNF-α)
  • Facilitates T cell activation
  • Enhances the TCR signaling pathway
  • Suitable for immunomodulation research
Catalog No. 50-003-47973 Supplier Medchemexpress LLC Supplier No. HY13633950MG
Cbl-b-IN-1 (Example 519) is an inhibitor of Cbl-b, with an IC50 of less than 100 nM. It is used in research on immunomodulation. It promotes the secretion of cytokines such as IL-2, IFN-γ, and TNF-α, facilitates T cell activation, and enhances the TCR signaling pathway.
  • Inhibitor of Cbl-b with an IC50 of less than 100 nM
  • Promotes secretion of cytokines (IL-2, IFN-γ, TNF-α)
  • Facilitates T cell activation
  • Enhances the TCR signaling pathway
  • Suitable for immunomodulation research