Learn More
Foxx Life Sciences EZwaste HPLC Solvent Waste System, Replacement Fittings, Six (6x) Blue Nut and Yellow Ferrule for 1/8 Inch OD, Six (6x) Black Plug for 1/8 Inch OD Port, Elbow and Straight Fittings for Carbon Filter, Black Plug for Carbon Filter Port, 1/PK
Catalog No. 501049968
Encompass_Preferred
330-0902-OEM: EZwaste Fittings, 1/8'' OD Fittings and 1/4'' Fittings Pack. Foxx Life Sciences' replacement fittings secure waste tubes to carbos, providing the closed system necessary to prevent spills. The EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety.
Catalog No. 50-104-9968 Supplier Foxx Life Sciences Supplier No. 3300902OEM
330-0902-OEM: EZwaste Fittings, 1/8'' OD Fittings and 1/4'' Fittings Pack. Foxx Life Sciences' replacement fittings secure waste tubes to carbos, providing the closed system necessary to prevent spills. The EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety.