Learn More
Foxx Life Sciences EZwaste HPLC Solvent Waste System, Cap-Only Assembly, 70mm (S70) VersaCap with Six (6x) Ports for 1/8 Inch O.D. Tubing; One (1x) Hose Barb Port for 1/4 or 3/8 Inch I.D. Tubing
Catalog No. 501049999
Encompass_Preferred
330-0B05-OEM: EZwaste System, UN/DOT, S-70 Cap, 6x 1/8" OD Tubing, 1x 1/4" or 1x 3/8" HB and Filter. Foxx Life Sciences EZwaste UN/DOT Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety. Each EZwaste UN/DOT Solvent Waste System comes with one S70 open Cap, fittings, plugs for unused ports, and a chemical exhaust filter. * Depending on application, carbon exhaust filter life cycle may vary. All UN/DOT containers are rated for transport for easy waste pickup.
Catalog No. 50-104-9999 Supplier Foxx Life Sciences Supplier No. 3300B05OEM
330-0B05-OEM: EZwaste System, UN/DOT, S-70 Cap, 6x 1/8" OD Tubing, 1x 1/4" or 1x 3/8" HB and Filter. Foxx Life Sciences EZwaste UN/DOT Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety. Each EZwaste UN/DOT Solvent Waste System comes with one S70 open Cap, fittings, plugs for unused ports, and a chemical exhaust filter. * Depending on application, carbon exhaust filter life cycle may vary. All UN/DOT containers are rated for transport for easy waste pickup.