Learn More
Cable Ties And More CBL TIES 11.6 IN 50LBS 100PCS
SDP
Catalog No. NC2752732
Encompass
Panduit Tefzel Zip Ties FeaturesMaterial- Ethylene-tetrafluoroethyleneHighly Resistant To Concentrated AcidsGood Radiation Resistance up to 100 MegaradsRoHS CompliantWeather ResistantUV Resistant: For Indoor or Outdoor useMax Operation Temperature 302 degree F / 150 degree CMeets the requirements of IEEE 383ASTM specification-UL83,ASTM-D2633,ASTM-D3159 Type 1
Catalog No. NC2752732 Supplier Cable Ties And More Supplier No. TF11
Panduit Tefzel Zip Ties FeaturesMaterial- Ethylene-tetrafluoroethyleneHighly Resistant To Concentrated AcidsGood Radiation Resistance up to 100 MegaradsRoHS CompliantWeather ResistantUV Resistant: For Indoor or Outdoor useMax Operation Temperature 302 degree F / 150 degree CMeets the requirements of IEEE 383ASTM specification-UL83,ASTM-D2633,ASTM-D3159 Type 1