Learn More
Medchemexpress LLC HY-18935A 2mg , CBL0137 hydrochloride CAS:1197397-89-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18935A2MG
Encompass_Preferred
HY-18935A 2mg CBL0137 hydrochloride CAS: 1197397-89-9 CBL0137 hydrochloride is an inhibitor of the histone chaperone, FACT. CBL0137 hydrochloride can also activate p53 and inhibits NF-κB with EC50s of 0.37 and 0.47 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1030