Learn More
Selleck Chemical LLC CBL0137 HCl
SDP
Catalog No. 5000002292
Encompass_Preferred
CBL0137 (CBLC137 Curaxin 137) HCl activates p53 and inhibits NF- B with EC50s of 0 37 M and 0 47 M in the cell-based p53 and NF-kB reporter assays respectively It also inhibits histone chaperone FACT (facilitates chromatin transcription complex)
Catalog No. 50-000-02292 Supplier Selleck Chemical LLC Supplier No. S8483100MG
Add to Cart
Add to Cart
CBL0137 (CBLC137 Curaxin 137) HCl activates p53 and inhibits NF- B with EC50s of 0 37 M and 0 47 M in the cell-based p53 and NF-kB reporter assays respectively It also inhibits histone chaperone FACT (facilitates chromatin transcription complex)