Learn More
TARGETMOL CHEMICALS INC CBL0137 HYDROCHLORIDE 5MG
SDP
Catalog No. 502576773
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CBL0137 hydrochloride (Curaxin-137 hydrochloride) activates p53 and inhibits NF-kB (EC50 0.37/0.47 uM) in the cell-based p53 and NF-kB reporter assays respectively. It also suppresses histone chaperone FACT. purity: 98%
Catalog No. 50-257-6773 Supplier TARGETMOL CHEMICALS INC Supplier No. T43615MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CBL0137 hydrochloride (Curaxin-137 hydrochloride) activates p53 and inhibits NF-kB (EC50 0.37/0.47 uM) in the cell-based p53 and NF-kB reporter assays respectively. It also suppresses histone chaperone FACT. purity: 98%