Learn More
Cayman Chemical CBL0137 (hydrochloride), Purity: ≥98%, CAS: 1197397-89-9, 10 mg
SDP
Catalog No. 502984538
Encompass_Preferred
A water soluble and metabolically stable curaxin that activates p53 with an EC50 value of 0.37 µM and inhibits NF-κB with an EC50 value of 0.47 µM; has broad anticancer activity in mice when administered orally
Catalog No. 50-298-4538 Supplier Cayman Chemical Supplier No. 1937310 MG
A water soluble and metabolically stable curaxin that activates p53 with an EC50 value of 0.37 µM and inhibits NF-κB with an EC50 value of 0.47 µM; has broad anticancer activity in mice when administered orally