Learn More
ACROBiosystems ACROBiosystems Human CBLB / RNF56 Protein, His Tag
Supplier:  ACROBiosystems CBBH51481MG
Encompass_Preferred
Recombinant Protein;1MG;Human CBLB, His Tag (CBB-H5148) is expressed from E.coli cells. It contains AA Ala 39 - Phe 426 (Accession # Q13191-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 47.0 kDa. The protein migrates as 46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6760