Learn More
Aviva Systems Biology Corp CBLL1 Antibody - N-terminal region (ARP39622_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39622_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CBLL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CBLL1. Alternate names: FLJ23109, HAKAI, MGC163401, MGC163403, RNF188ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57114