Learn More
ABclonal Technology CBLL1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14932500UL
Encompass_Preferred
This gene encodes an E3 ubiquitin-ligase for the E-cadherin complex and mediates its ubiquitination, endocytosis, and degradation in the lysosomes. The encoded protein contains a RING-finger domain and is also thought to have a role in control of cell proliferation. A related pseudogene has been identified on chromosome X. Alternative splicing results in a non-coding transcript variant.
Catalog No. 50-001-05274