Learn More
Lonza Walkersville CBM Chondrocyte Growth Basal Medium
Catalog No. NC1925834
Encompass
CBM is a Chondrocyte cell growth basal medium sold separately. This can be used with 1 kit of CC-4409 (chondrocyte cell growth medium SingleQuots Supplements and Growth Factors). The full CGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3216.CGM Chondrocyte Growth Medium is specially designed for expansion of chondrocytes. The BulletKit is conveniently packaged with basal medium and supplements included for the growth of chondrocytes.CDM Chondrocyte Differentiation Medium is also available and sold separately for differentiation of human chondrocytesContent:1 x Bottle Basal Medium 500 mL
Catalog No. NC1925834 Supplier Lonza Walkersville Supplier No. CC3217
CBM is a Chondrocyte cell growth basal medium sold separately. This can be used with 1 kit of CC-4409 (chondrocyte cell growth medium SingleQuots Supplements and Growth Factors). The full CGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3216.CGM Chondrocyte Growth Medium is specially designed for expansion of chondrocytes. The BulletKit is conveniently packaged with basal medium and supplements included for the growth of chondrocytes.CDM Chondrocyte Differentiation Medium is also available and sold separately for differentiation of human chondrocytesContent:1 x Bottle Basal Medium 500 mL