Learn More
Foxx Life Sciences Magnetic Stir Bar Mixer, 55mm Length, 8mm Diameter, Double Ended Dumbell Shape
Catalog No. 501050017
Encompass_Preferred
257-1101-OEM: Magnetic Mixer, Double Ended, 8mm Diameter, 55mm Long. Foxx Life Sciences Magnetic Mixers are used to mix components in order to obtain a homogeneous liquid mixtures. Our mixers can be used in a variety of applications including but not limited to chemistry and biology experiments, buffer solutions, and bacterial growth media. Foxx magnetic mixers are ideal for mixing over a long period of time. Since our mixers are just magnet bars, they can easily be cleaned and the risks of contaminating the solution can be minimized.
Catalog No. 50-105-0017 Supplier Foxx Life Sciences Supplier No. 2571101OEM
257-1101-OEM: Magnetic Mixer, Double Ended, 8mm Diameter, 55mm Long. Foxx Life Sciences Magnetic Mixers are used to mix components in order to obtain a homogeneous liquid mixtures. Our mixers can be used in a variety of applications including but not limited to chemistry and biology experiments, buffer solutions, and bacterial growth media. Foxx magnetic mixers are ideal for mixing over a long period of time. Since our mixers are just magnet bars, they can easily be cleaned and the risks of contaminating the solution can be minimized.