Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CBP-501 acetate | 1829512-40-4 | 99.8% | 1,989.1 g/mol | C88H126F5N29O19 | 10 MG
SDP

Catalog No. 5000288795
Encompass_Preferred

CBP-501 acetate is a cell-permeable calmodulin-binding peptide supplied as a powder for research use. It acts as a G2 checkpoint abrogator, sensitizes cells to DNA-damaging agents, and inhibits multiple Ser216-specific kinases.

  • Cell-permeable calmodulin-binding peptide.
  • G2 checkpoint abrogator that sensitizes cells to DNA damage.
  • Inhibits multiple Ser216-specific kinases such as CHK1 and CHK2.
  • Peptide sequence: d({4-Bz-Phe}-Ser-Trp-Ser-{pentafluoro-Phe}-{Cha}-Arg-Arg-Arg-Gln-Arg-Arg).
  • Physical form: powder; sealed storage away from moisture, store at -80°C or -20°C per handling instructions.
  • High purity (~99.8%) suitable for research applications.

Catalog No. 50-002-88795 Supplier Medchemexpress LLC Supplier No. HY16129A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CBP-501 acetate is a cell-permeable calmodulin-binding peptide supplied as a powder for research use. It acts as a G2 checkpoint abrogator, sensitizes cells to DNA-damaging agents, and inhibits multiple Ser216-specific kinases.

  • Cell-permeable calmodulin-binding peptide.
  • G2 checkpoint abrogator that sensitizes cells to DNA damage.
  • Inhibits multiple Ser216-specific kinases such as CHK1 and CHK2.
  • Peptide sequence: d({4-Bz-Phe}-Ser-Trp-Ser-{pentafluoro-Phe}-{Cha}-Arg-Arg-Arg-Gln-Arg-Arg).
  • Physical form: powder; sealed storage away from moisture, store at -80°C or -20°C per handling instructions.
  • High purity (~99.8%) suitable for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.