Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CBP-501 (acetate) | 1929.06 (free base) | 1 MG
SDP

Catalog No. 5000392882
Encompass_Preferred

CBP-501 acetate is a cell-permeable calmodulin-binding peptide and a G2-abrogating drug candidate. It inhibits the activity of multiple Ser216-specific kinases, including MAPKAP-K2, C-Tak1, CHK1, and CHK2. It is used for various types of cancer and enhances the cytotoxicity of CDDP and Bleomycin against cancer cells both in vitro and in vivo.

  • Inhibits MAPKAP-K2 with an IC50 of 0.9 μM
  • Inhibits C-Tak1 with an IC50 of 1.4 μM
  • Inhibits Chk1 with an IC50 of 3.4 μM
  • Inhibits Chk2 with an IC50 of 6.5 μM
  • Stable for 2 years at -80°C as powder
  • Stable for 1 year at -20°C as powder
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent
  • Enhances cytotoxicity of CDDP and Bleomycin

Catalog No. 50-003-92882 Supplier Medchemexpress LLC Supplier No. HY16129A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CBP-501 acetate is a cell-permeable calmodulin-binding peptide and a G2-abrogating drug candidate. It inhibits the activity of multiple Ser216-specific kinases, including MAPKAP-K2, C-Tak1, CHK1, and CHK2. It is used for various types of cancer and enhances the cytotoxicity of CDDP and Bleomycin against cancer cells both in vitro and in vivo.

  • Inhibits MAPKAP-K2 with an IC50 of 0.9 μM
  • Inhibits C-Tak1 with an IC50 of 1.4 μM
  • Inhibits Chk1 with an IC50 of 3.4 μM
  • Inhibits Chk2 with an IC50 of 6.5 μM
  • Stable for 2 years at -80°C as powder
  • Stable for 1 year at -20°C as powder
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent
  • Enhances cytotoxicity of CDDP and Bleomycin

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.