Learn More
Medchemexpress LLC HY-111784 10mg Medchemexpress, CBP-IN-1 CAS:2222941-37-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11178410MG
Encompass_Preferred
Medchemexpress, HY-111784 10mg CBP-IN-1 CAS:2222941-37-7 CBP-IN-1 is a potent p300/CBP bromodomain inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8962