Learn More
Medchemexpress LLC HY-111784 5mg Medchemexpress, CBP-IN-1 CAS:2222941-37-7 Purity:>98%
SDP
Catalog No. 501932394
Encompass_Preferred
Medchemexpress, HY-111784 5mg CBP-IN-1 CAS:2222941-37-7 CBP-IN-1 is a potent p300 / CBP bromodomain inhibitor [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2394 Supplier Medchemexpress LLC Supplier No. HY1117845MG
Medchemexpress, HY-111784 5mg CBP-IN-1 CAS:2222941-37-7 CBP-IN-1 is a potent p300 / CBP bromodomain inhibitor [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.