Learn More
Medchemexpress LLC Cbp/p300-in-19 hydrochloride | 2592638-14-5 | 99.00% | 514.01 | 1 ML
SDP
Catalog No. 5000363434
Encompass_Preferred
CBP/p300-IN-19 hydrochloride is a potent and selective p300/CBP HAT inhibitor that also shows antitumor activity.
  • Potent and selective p300/CBP HAT inhibitor.
  • IC50 for p300-HAT is 1.4 μM.
  • IC50 for CBP-HAT is 2.2 μM.
  • IC50 for PCAF is >100 μM.
  • IC50 for Myst3 is >100 μM.
  • Shows antitumor activity.
Catalog No. 50-003-63434 Supplier Medchemexpress LLC Supplier No. HY146277A10MM/1ML
CBP/p300-IN-19 hydrochloride is a potent and selective p300/CBP HAT inhibitor that also shows antitumor activity.
  • Potent and selective p300/CBP HAT inhibitor.
  • IC50 for p300-HAT is 1.4 μM.
  • IC50 for CBP-HAT is 2.2 μM.
  • IC50 for PCAF is >100 μM.
  • IC50 for Myst3 is >100 μM.
  • Shows antitumor activity.