Learn More
Medchemexpress LLC Cbp/p300-in-3 | 2299226-01-8 | 98.0% | 431.53 g/mol | C24H29N7O | 5 MG
SDP
Catalog No. 5000262876
Encompass_Preferred
CBP/p300-IN-3 is a small-molecule inhibitor of the p300/CBP histone acetyltransferases (HATs), described as Compound 6 in patent WO 2019049061 A1. The compound is supplied for research use and is accompanied by analytical documentation and safety information.
  • Small-molecule inhibitor of p300/CBP histone acetyltransferases.
  • High chemical purity (98.0%).
  • Molecular formula C24H29N7O and molecular weight 431.53 g/mol.
  • Available in multiple pack sizes, including 5 mg.
  • Accompanied by COA, SDS, and data sheet documentation.
  • Intended for research use only.
Catalog No. 50-002-62876 Supplier Medchemexpress LLC Supplier No. HY1288765MG
CBP/p300-IN-3 is a small-molecule inhibitor of the p300/CBP histone acetyltransferases (HATs), described as Compound 6 in patent WO 2019049061 A1. The compound is supplied for research use and is accompanied by analytical documentation and safety information.
  • Small-molecule inhibitor of p300/CBP histone acetyltransferases.
  • High chemical purity (98.0%).
  • Molecular formula C24H29N7O and molecular weight 431.53 g/mol.
  • Available in multiple pack sizes, including 5 mg.
  • Accompanied by COA, SDS, and data sheet documentation.
  • Intended for research use only.