Learn More
Medchemexpress LLC CBP/p300-IN-3 | 2299226-01-8 | 98.01% | 100 MG
SDP
Catalog No. 5000341186
Encompass_Preferred
CBP/p300-IN-3 is a p300/CBP histone acetyltransferase inhibitor, identified as Compound 6, and is sourced from patent WO 2019049061 A1.
  • p300/CBP histone acetyltransferase inhibitor.
  • Identified as Compound 6.
  • Sourced from patent WO 2019049061 A1.
  • For research use only.
  • Not sold to patients.
Catalog No. 50-003-41186 Supplier Medchemexpress LLC Supplier No. HY128876100MG
CBP/p300-IN-3 is a p300/CBP histone acetyltransferase inhibitor, identified as Compound 6, and is sourced from patent WO 2019049061 A1.
  • p300/CBP histone acetyltransferase inhibitor.
  • Identified as Compound 6.
  • Sourced from patent WO 2019049061 A1.
  • For research use only.
  • Not sold to patients.