Learn More
Medchemexpress LLC CBP/p300-IN-8 | 2304416-91-7 | 99.9% | 461.55 | 50 MG
SDP
Catalog No. 5000348772
Encompass_Preferred
CBP/p300-IN-8 is a potent inhibitor of the CBP/P300 family of bromodomains. It inhibits CBP activity with an IC50 of 0.01-0.1 μM and BRD4 activity with an IC50 of 1-1000 μM.
  • Potent inhibitor of the CBP/P300 family of bromodomains.
  • Inhibits CBP activity with an IC50 of 0.01-0.1 μM.
  • Inhibits BRD4 activity with an IC50 of 1-1000 μM.
Catalog No. 50-003-48772 Supplier Medchemexpress LLC Supplier No. HY13692050MG
CBP/p300-IN-8 is a potent inhibitor of the CBP/P300 family of bromodomains. It inhibits CBP activity with an IC50 of 0.01-0.1 μM and BRD4 activity with an IC50 of 1-1000 μM.
  • Potent inhibitor of the CBP/P300 family of bromodomains.
  • Inhibits CBP activity with an IC50 of 0.01-0.1 μM.
  • Inhibits BRD4 activity with an IC50 of 1-1000 μM.