Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cbp/p300-in-8 | 2304416-91-7 | 99.9% | C27H31N3O4 | 10MG
SDP

Catalog No. 5000260894
Encompass_Preferred

CBP/p300-IN-8 is a small-molecule inhibitor that targets the bromodomains of the CBP/p300 family and is used in epigenetic research. It inhibits CBP at low-nanomolar concentrations and exhibits activity against BRD4, and is supplied as a high-purity solid reagent for laboratory studies.

  • Potent inhibitor of CBP/p300 bromodomains.
  • CBP IC50 0.01-0.1 μM; BRD4 IC50 1-1000 μM.
  • Molecular formula C27H31N3O4; molecular weight 461.55.
  • High purity (99.94%).
  • White to off-white solid for analytical and biological assays.
  • Available in small research pack sizes with room-temperature shipping options.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-60894 Supplier Medchemexpress LLC Supplier No. HY13692010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CBP/p300-IN-8 is a small-molecule inhibitor that targets the bromodomains of the CBP/p300 family and is used in epigenetic research. It inhibits CBP at low-nanomolar concentrations and exhibits activity against BRD4, and is supplied as a high-purity solid reagent for laboratory studies.

  • Potent inhibitor of CBP/p300 bromodomains.
  • CBP IC50 0.01-0.1 μM; BRD4 IC50 1-1000 μM.
  • Molecular formula C27H31N3O4; molecular weight 461.55.
  • High purity (99.94%).
  • White to off-white solid for analytical and biological assays.
  • Available in small research pack sizes with room-temperature shipping options.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.