Learn More
Medchemexpress LLC HY-136920 5mg Medchemexpress, CBP/p300-IN-8 CAS:2304416-91-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1369205MG
Encompass_Preferred
Medchemexpress, HY-136920 5mg CBP/p300-IN-8 CAS:2304416-91-7 CBP/p300-IN-8 is a potent inhibitor of the CBP/P300 family of bromodomains. CBP/p300-IN-8 inhibits CBP (IC50=0.01-0.1 µΜ) and BRD4 (IC50=1-1000 µΜ) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1465