Learn More
ABclonal Technology CBP80 Rabbit pAb
SDP
Supplier:  ABclonal Technology A139391000UL
Encompass_Preferred
The product of this gene is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5 cap of nascent pre-mRNA in the nucleoplasm. The encoded protein promotes high-affinity mRNA-cap binding and associates with the CTD of RNA polymerase II. The CBC promotes pre-mRNA splicing, 3-end processing, RNA nuclear export, and nonsense-mediated mRNA decay.
Catalog No. 50-001-09011