Learn More
Medchemexpress LLC CBPD-409, a PROTAC CBP/p300 degrader | 3037549-86-0 | 99.4% | 862.97 g·mol⁻¹ | C46H52F2N10O5 | 10 MG
SDP
Catalog No. 5000288425
Encompass_Preferred
CBPD-409 is an orally active PROTAC degrader that targets the CBP and p300 bromodomain-containing proteins. It shows potent degradation (DC50 0.2-0.4 nM) and low-nanomolar antiproliferative activity in AR+ prostate cancer cell lines, making it suitable for preclinical studies of CBP/p300 biology and degrader pharmacology.
  • Potent degradation activity (DC50 0.2-0.4 nM)
  • Low-nanomolar antiproliferative activity in AR+ prostate cancer cell lines (IC50 1.2-2.0 nM)
  • High supplied purity (>99%)
  • Supplied as a powder suitable for formulation in solvent for assays
  • Recommended storage to preserve stability: powder -20°C long term; in solvent -80°C up to 6 months
Catalog No. 50-002-88425 Supplier Medchemexpress LLC Supplier No. HY15811310MG
CBPD-409 is an orally active PROTAC degrader that targets the CBP and p300 bromodomain-containing proteins. It shows potent degradation (DC50 0.2-0.4 nM) and low-nanomolar antiproliferative activity in AR+ prostate cancer cell lines, making it suitable for preclinical studies of CBP/p300 biology and degrader pharmacology.
  • Potent degradation activity (DC50 0.2-0.4 nM)
  • Low-nanomolar antiproliferative activity in AR+ prostate cancer cell lines (IC50 1.2-2.0 nM)
  • High supplied purity (>99%)
  • Supplied as a powder suitable for formulation in solvent for assays
  • Recommended storage to preserve stability: powder -20°C long term; in solvent -80°C up to 6 months