Learn More
Medchemexpress LLC HY-134205A 5mg Medchemexpress, CBR-470-1 CAS:2416095-06-0 Purity:>98%
SDP
Catalog No. 502081359
Encompass_Preferred
Medchemexpress, HY-134205A 5mg CBR-470-1 CAS:2416095-06-0 CBR-470-1 is an inhibitor of the glycolytic enzyme phosphoglycerate kinase 1 (PGK1). CBR-470-1 is also a non-covalent Nrf2 activator. CBR-470-1 protects SH-SY5Y neuronal cells against MPP+-induced cytotoxicity through activation of the Keap1-Nrf2 cascade. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1359 Supplier Medchemexpress LLC Supplier No. HY134205A5MG
Medchemexpress, HY-134205A 5mg CBR-470-1 CAS:2416095-06-0 CBR-470-1 is an inhibitor of the glycolytic enzyme phosphoglycerate kinase 1 (PGK1). CBR-470-1 is also a non-covalent Nrf2 activator. CBR-470-1 protects SH-SY5Y neuronal cells against MPP+-induced cytotoxicity through activation of the Keap1-Nrf2 cascade. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.