Learn More
Aviva Systems Biology Corp CBR1 Antibody - C-terminal region (ARP45801_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45801_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CBR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CBR1. Alternate names: CBR, hCBR1, SDR21C1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53999