Learn More
Aviva Systems Biology Corp CBR4 Antibody - N-terminal region (ARP60665_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60665_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CBR4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CBR4. Alternate names: FLJ14431, SDR45C1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-121