Learn More
Medchemexpress LLC HY-131059 5mg Medchemexpress, CBS1117 CAS:959245-08-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1310595MG
Encompass_Preferred
Medchemexpress, HY-131059 5mg CBS1117 CAS:959245-08-0 CBS1117 is a virus entry inhibitor with an IC50 of 70 nM for influenza A virus, A/Puerto Rico/8/34 (H1N1). CBS1117 interferes with the hemagglutinin (HA)-mediated fusion process. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1261