Learn More
Adam Equipment Adam Equipment CBX 2001 Compact Balance 3000g Capacity x 0.5g Readability
Catalog No. NC2021382
Encompass_Preferred
Adam Equipment CBX balances are powered by a rechargeable battery and can be recharged on-the-go via the USB interface. Solid ABS housing and a grade 304 stainless steel ensure the CBX is easy to clean even during demanding field operation. Parts counting is easy with selectable preset sample sizes. The bright backlit display, which is highly visible for clear results in most lighting conditions, features 18mm-high digits and a low-battery indicator. Levelling feet and a bubble indicator simplify setup to ensure quick, reliable results anywhere.
Catalog No. NC2021382 Supplier Adam Equipment Supplier No. CBX 3001
Adam Equipment CBX balances are powered by a rechargeable battery and can be recharged on-the-go via the USB interface. Solid ABS housing and a grade 304 stainless steel ensure the CBX is easy to clean even during demanding field operation. Parts counting is easy with selectable preset sample sizes. The bright backlit display, which is highly visible for clear results in most lighting conditions, features 18mm-high digits and a low-battery indicator. Levelling feet and a bubble indicator simplify setup to ensure quick, reliable results anywhere.