Learn More
Aviva Systems Biology Corp CBX1 Antibody - middle region (ARP34427_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34427_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CBX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CBX1. Alternate names: CBX, M31, MOD1, p25beta, HP1-BETA, HP1Hsbeta, HP1Hs-betaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55389