Learn More
Aviva Systems Biology Corp CBX1 Antibody - N-terminal region (ARP34426_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34426_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CBX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CBX1. Alternate names: CBX, HP1-BETA, HP1Hs-beta, M31, MOD1, p25beta, HP1HsbetaApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-55388