Learn More
ABclonal Technology CBX1/HP1 beta Rabbit mAb
SDP
Catalog No. 5000005151
Encompass_Preferred
This gene encodes a highly conserved nonhistone protein which is a member of the heterochromatin protein family The protein is enriched in the heterochromatin and associated with centromeres The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues and a C-terminal chromo shadow-domain (CSD) which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins The protein may play an important role in the epigenetic control of chromatin structure and gene expression Several related pseudogenes are located on chromosomes 1 3 and X Multiple alternatively spliced variants encoding the same protein have been identified
Catalog No. 50-000-05151 Supplier ABclonal Technology Supplier No. A2270820UL
This gene encodes a highly conserved nonhistone protein which is a member of the heterochromatin protein family The protein is enriched in the heterochromatin and associated with centromeres The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues and a C-terminal chromo shadow-domain (CSD) which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins The protein may play an important role in the epigenetic control of chromatin structure and gene expression Several related pseudogenes are located on chromosomes 1 3 and X Multiple alternatively spliced variants encoding the same protein have been identified