Learn More
Aviva Systems Biology Corp CBX2 Antibody - N-terminal region (ARP51628_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51628_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CBX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CBX2. Alternate names: CDCA6, M33, MGC10561, SRXY5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53998