Learn More
Aviva Systems Biology Corp CBX2 Antibody - middle region (ARP58370_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58370_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CBX2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CBX2. Alternate names: CDCA6, M33, MGC10561, SRXY5ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rat
Catalog No. 50-286-469