Learn More
Cayman Chemical CBX2 chromodomaInhumn reco
SDP
Catalog No. 502971024
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • MW: 38.2 kDa
Catalog No. 50-297-1024 Supplier Cayman Chemical Supplier No. 14769100 UG
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • MW: 38.2 kDa