Learn More
Aviva Systems Biology Corp CBX3 Antibody - middle region (ARP34721_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34721_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CBX3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human CBX3. Alternate names: HECH, HP1-GAMMA, HP1Hs-gammaApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55532