Learn More
Aviva Systems Biology Corp CBX4 Antibody - N-terminal region (ARP30002_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30002_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CBX4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CBX4. Alternate names: PC2, NBP16ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54092