Learn More
Cayman Chemical CBX5humn recombInt 100ug
SDP
Catalog No. 502951412
Encompass_Preferred
Source: Recombinant N-terminal His-tagged protein expressed in E. coli • MW: 24.1 kDa • Amino acids: 2-191 (full length)
Catalog No. 50-295-1412 Supplier Cayman Chemical Supplier No. 11235100 UG
Source: Recombinant N-terminal His-tagged protein expressed in E. coli • MW: 24.1 kDa • Amino acids: 2-191 (full length)