Learn More
Aviva Systems Biology Corp CBX6 Antibody - C-terminal region (ARP39074_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39074_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CBX6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CBX6. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56900